Gene Transcripts

Gene Symbol
Ifrd1
Entrez Gene ID
29596
Gene Name
interferon-related developmental regulator 1
Gene Aliases
Pc4
Chromosome Location
Chr. 6: 60132018 - 60151354 on Build Rnor_6.0
UniGene ID
Rn.3723
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019242.1 NP_062115.1 2 282
XM_008764643.1 XP_008762865.1 1 901
GenBank mRNA BC083560.1 - 2 348
J04511.1 - 2 282

View Full Product Details