Gene Transcripts

Gene Symbol
Sh2b3
Entrez Gene ID
58838
Gene Name
SH2B adaptor protein 3
Gene Aliases
Lnk
Chromosome Location
Chr. 12: 40261990 - 40265757 on Build Rnor_6.0
UniGene ID
Rn.11228
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031621.1 NP_113809.1 2 1861
GenBank mRNA JX215366.1 - 2495
U24652.1 - 6 1466
U24653.1 - 6 1517
U24654.1 - 5 1597
U24655.1 - 2 1861

View Full Product Details