Gene Transcripts

Gene Symbol
Grik4
Entrez Gene ID
24406
Gene Name
glutamate receptor, ionotropic, kainate 4
Gene Aliases
GluK4, KA1
Chromosome Location
Chr. 8: 46806147 - 47107917 on Build Rnor_6.0
UniGene ID
Rn.10049
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012572.1 NP_036704.1 13 1603
GenBank mRNA U08257.1 - 13 1603
X59996.1 - 13 1604

View Full Product Details