Gene Transcripts

Gene Symbol
Pfkfb1
Entrez Gene ID
24638
Gene Name
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 1
Gene Aliases
PFRX, Pfkfb01
Chromosome Location
Chr. X: 23092143 - 23145919 on Build Rnor_6.0
UniGene ID
Rn.10115
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA J04197.1 - 5, 6 654

View Full Product Details