Gene Transcripts

Gene Symbol
Ren
Entrez Gene ID
24715
Gene Name
renin
Gene Aliases
RATRENAA, RENAA, Ren1
Chromosome Location
Chr. 13: 50502724 - 50513953 on Build Rnor_6.0
UniGene ID
Rn.9831
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012642.4 NP_036774.4 1, 2 113
GenBank mRNA BC078878.1 - 1, 2 113
J02941.1 - 1, 2 306
S60054.1 - 1, 2 123
X07033.1 - 1, 2 123

View Full Product Details