Gene Transcripts
- Gene Symbol
- P2ry1
- Entrez Gene ID
- 25265
- Gene Name
- purinergic receptor P2Y, G-protein coupled, 1
- Gene Aliases
- P2y, P2y1
- Chromosome Location
- Chr. 2: 151314818 - 151320312 on Build Rnor_6.0
- UniGene ID
- Rn.10217
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_012800.1 |
NP_036932.1
|
1 |
1354 |
|
GenBank mRNA |
U22830.1 |
- |
1 |
1354 |
|
View Full Product Details