Gene Transcripts

Gene Symbol
Adh1
Entrez Gene ID
24172
Gene Name
alcohol dehydrogenase 1 (class I)
Gene Aliases
Adh, Adh1a, Adh1c
Chromosome Location
Chr. 2: 243550657 - 243562243 on Build Rnor_6.0
UniGene ID
Rn.40222
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF508795.1 - 4 416
BC062403.1 - 4 388 5621394
CK480694.1 - 4 381
FQ209590.1 - 4 399
FQ218571.1 - 4 401
FQ220674.1 - 4 399
M15327.1 - 4 372
X72792.1 - 2 202

View Full Product Details