Gene Transcripts

Gene Symbol
Qrfpr
Entrez Gene ID
310327
Gene Name
pyroglutamylated RFamide peptide receptor
Gene Aliases
AQ27, Gpr103
Chromosome Location
Chr. 2: 122891321 - 122949241 on Build Rnor_6.0
UniGene ID
Rn.162584
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198199.1 NP_937842.1 1 77
GenBank mRNA AB109630.1 - 1 77

View Full Product Details