Gene Transcripts

Gene Symbol
Cyp24a1
Entrez Gene ID
25279
Gene Name
cytochrome P450, family 24, subfamily a, polypeptide 1
Gene Aliases
25-hydroxyvitaminD324-hydroxylase, Cyp24, VITD12
Chromosome Location
Chr. 3: 168097484 - 168111917 on Build Rnor_6.0
UniGene ID
Rn.100353
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_201635.2 NP_963966.1 8 1123
XM_006235672.2 XP_006235734.1 8 1465
GenBank mRNA BC100059.1 - 8 1468
X59506.1 - 8 1466

View Full Product Details