Gene Transcripts

Gene Symbol
Ube4a
Entrez Gene ID
315608
Gene Name
ubiquitination factor E4A
Gene Aliases
Ab2-232
Chromosome Location
Chr. 8: 49229713 - 49271896 on Build Rnor_6.0
UniGene ID
Rn.102204
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207610.1 NP_997493.1 2 255
XM_006242938.2 XP_006243000.1 3 412
GenBank mRNA AY325198.1 - 2 150
BC061761.1 - 2 255 5621078

View Full Product Details