Gene Transcripts

Gene Symbol
MKNK1
Entrez Gene ID
8569
Gene Name
MAP kinase interacting serine/threonine kinase 1
Gene Aliases
MNK1
Chromosome Location
Chr. 1: 46557407 - 46607140 on Build GRCh38
UniGene ID
Hs.371594
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135553.2 NP_001129025.1 4 427
NM_003684.5 NP_003675.2 4 427
NM_198973.3 NP_945324.1 4 385
NR_024174.2 4 427
NR_024176.2 5 528
XM_006711000.1 XP_006711063.1 4 380
XM_006711001.2 XP_006711064.1 5 630
XM_006711002.2 XP_006711065.1 4 5855
XM_006711003.1 XP_006711066.1 5 441
XM_006711013.1 XP_006711076.1 4 379
XM_006711014.1 XP_006711077.1 4 370
XM_006711015.1 XP_006711078.1 5 441
XM_017002655.1 XP_016858144.1 5 6201
XM_017002656.1 XP_016858145.1 5 6000
XM_017002659.1 XP_016858148.1 4 399
XM_017002660.1 XP_016858149.1 4 5855
XM_017002661.1 XP_016858150.1 5 620
XM_017002662.1 XP_016858151.1 5 441
XM_017002663.1 XP_016858152.1 5 6201
XM_017002664.1 XP_016858153.1 4 5855
XM_017002666.1 XP_016858155.1 5 3330
XM_017002667.1 XP_016858156.1 3 906
GenBank mRNA AB000409.1 - 4 370
AB451300.1 - 3 183
AB451432.1 - 3 183
AK096423.1 - 5 472
AK290456.1 - 4 347
AK298839.1 - 4 401
AK304005.1 - 4 450
AY355461.1 - 3 183
BC002755.2 - 4 347 3629765
BG722049.1 - 4 390
BX538193.1 - 3 251
BX648559.1 - 5 436
DA435648.1 - 4 372
DA502363.1 - 4 400
DC366212.1 - 4 401
DC376088.1 - 4 357

View Full Product Details