Gene Transcripts

Gene Symbol
ADGRE2
Entrez Gene ID
30817
Gene Name
adhesion G protein-coupled receptor E2
Gene Aliases
CD312, EMR2, VBU
Chromosome Location
Chr. 19: 14732697 - 14778541 on Build GRCh38
UniGene ID
Hs.531619
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271052.1 NP_001257981.1 9 1062
NM_013447.3 NP_038475.2 10 1347
XM_011527948.2 XP_011526250.1 9 976
XM_011527949.2 XP_011526251.1 9 976
XM_011527951.2 XP_011526253.1 8 829
XM_011527952.2 XP_011526254.1 9 976
XM_011527953.2 XP_011526255.1 7 783
XM_011527954.2 XP_011526256.1 6 1514
XM_011527955.2 XP_011526257.1 4 393
XM_017026726.1 XP_016882215.1 8 945
XM_017026727.1 XP_016882216.1 8 829
GenBank mRNA AF114491.1 - 9 960
AK024426.1 - 2 114
AK222891.1 - 9 978
AK291776.1 - 9 1093
AK298690.1 - 5 422
AK298700.1 - 9 978
AK301458.1 - 8 829
BC034949.1 - 9 1782
BC053933.1 - 9 1775
BC082991.1 - 9 937
BC127004.1 - 10 1343 40108332
BC127005.1 - 10 1343 40108333
CR936819.1 - 4 357

View Full Product Details