Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014026.4 | NP_054745.1 | 1 | 415 | |
XM_011542778.2 | XP_011541080.1 | 1 | 123 | ||
XM_017017612.1 | XP_016873101.1 | 1 | 866 | ||
GenBank mRNA | AF077201.1 | - | 1 | 98 | |
AF532613.1 | - | 1 | 415 | ||
AK026554.1 | - | 1 | 111 | ||
AK223089.1 | - | 1 | 111 | ||
AK307403.1 | - | 1 | 107 | ||
AY040771.1 | - | 1 | 113 | ||
AY077684.1 | - | 1 | 86 | ||
BC014532.1 | - | 1 | 108 | 4064215 | |
BE382749.1 | - | 1 | 77 |