Gene Transcripts

Gene Symbol
C11orf54
Entrez Gene ID
28970
Gene Name
chromosome 11 open reading frame 54
Gene Aliases
PTD012, PTOD012
Chromosome Location
Chr. 11: 93741594 - 93765323 on Build GRCh38
UniGene ID
Hs.8360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286067.1 NP_001272996.1 10 1775
NM_001286068.1 NP_001272997.1 9 1638
NM_001286069.1 NP_001272998.1 9 1643
NM_001286070.1 NP_001272999.1 8 1491
NM_001286071.1 NP_001273000.1 8 1466
NM_014039.3 NP_054758.2 8 1493
XM_006718824.1 XP_006718887.1 10 1684
XM_011542781.2 XP_011541083.1 9 1957
XM_011542782.2 XP_011541084.1 9 2048
XM_017017613.1 XP_016873102.1 10 5099
XM_017017614.1 XP_016873103.1 10 1926
XM_017017615.1 XP_016873104.1 10 2379
XM_017017616.1 XP_016873105.1 8 1485
XM_017017617.1 XP_016873106.1 8 1881
XM_017017618.1 XP_016873107.1 8 1457
XM_017017619.1 XP_016873108.1 7 1339
GenBank mRNA AK129932.1 - 4 1060
AK292215.1 - 10 1694
AY203960.1 - 8 1518
BC007110.1 - 8 1398
BC012298.1 - 8 1460 3852558

View Full Product Details