Gene Transcripts

Gene Symbol
TENM1
Entrez Gene ID
10178
Gene Name
teneurin transmembrane protein 1
Gene Aliases
ODZ1, ODZ3, TEN-M1, TNM, TNM1
Chromosome Location
Chr. X: 124374680 - 125204305 on Build GRCh38
UniGene ID
Hs.23796
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163278.1 NP_001156750.1 1 233
NM_001163279.1 NP_001156751.1 1 233
NM_014253.3 NP_055068.2 1 233
XM_011531230.2 XP_011529532.1 4 874
XM_017029208.1 XP_016884697.1 4 874
XM_017029209.1 XP_016884698.1 5 929
XM_017029210.1 XP_016884699.1 3 802
XM_017029211.1 XP_016884700.1 4 880
XM_017029212.1 XP_016884701.1 4 491
XM_017029213.1 XP_016884702.1 3 803
XM_017029214.1 XP_016884703.1 4 874
XM_017029215.1 XP_016884704.1 4 873
XM_017029216.1 XP_016884705.1 4 873
GenBank mRNA AF100772.1 - 1 233
BC140783.1 - 1 230 9021651
BC144543.1 - 1 230 9053070
HE578281.1 - 1 169

View Full Product Details