Gene Transcripts

Gene Symbol
PATZ1
Entrez Gene ID
23598
Gene Name
POZ/BTB and AT hook containing zinc finger 1
Gene Aliases
MAZR, PATZ, RIAZ, ZBTB19, ZNF278, ZSG, dJ400N23
Chromosome Location
Chr. 22: 31325804 - 31346263 on Build GRCh38
UniGene ID
Hs.731398
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014323.2 NP_055138.2 1 1128
NM_032050.1 NP_114439.1 1 1128
NM_032051.1 NP_114440.1 1 1128
NM_032052.1 NP_114441.1 1 1128
GenBank mRNA AB208787.1 - 2 498
AF119256.1 - 1 1127
AF242522.1 - 1 484
AF254082.1 - 1 1097
AF254083.1 - 1 1097
AF254084.1 - 1 1097
AF254085.1 - 1 1097
AK291803.1 - 1 641
AL096880.1 - 1 462
AY028384.1 - 1 858
BC021091.1 - 1 675 4857537
CR456613.1 - 1 566

View Full Product Details