Gene Transcripts

Gene Symbol
RAB26
Entrez Gene ID
25837
Gene Name
RAB26, member RAS oncogene family
Gene Aliases
V46133
Chromosome Location
Chr. 16: 2148144 - 2154165 on Build GRCh38
UniGene ID
Hs.3797
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308053.1 NP_001294982.1 8, 9 772
NM_014353.4 NP_055168.2 7, 8 733
XM_011522450.2 XP_011520752.1 6, 7 1550
GenBank mRNA AB027137.1 - 6, 7 404
AF498952.1 - 6, 7 401
AK314110.1 - 8, 9 772
AY646153.1 - 7, 8 750
BC066913.1 - 7, 8 739 5262795

View Full Product Details