Gene Transcripts

Gene Symbol
HIST1H3B
Entrez Gene ID
8358
Gene Name
histone cluster 1, H3b
Gene Aliases
H3/l, H3FL
Chromosome Location
Chr. 6: 26031589 - 26032060 on Build GRCh38
UniGene ID
Hs.533292
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003537.3 NP_003528.1 1 184
GenBank mRNA BC069133.1 - 1 184
BC096132.1 - 1 184 40002522
BC096133.3 - 1 184 40002523
BC096134.3 - 1 184 40002524
BC099630.3 - 1 187 40002521

View Full Product Details