Gene Transcripts

Gene Symbol
MCHR1
Entrez Gene ID
2847
Gene Name
melanin concentrating hormone receptor 1
Gene Aliases
GPR24, MCH-1R, MCH1R, SLC-1, SLC1
Chromosome Location
Chr. 22: 40679178 - 40682814 on Build GRCh38
UniGene ID
Hs.248122
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005297.3 NP_005288.3 2 2257
GenBank mRNA AF490537.2 - 2 2340

View Full Product Details