Gene Transcripts

Gene Symbol
CPNE7
Entrez Gene ID
27132
Gene Name
copine 7
Gene Aliases
Chromosome Location
Chr. 16: 89575768 - 89597246 on Build GRCh38
UniGene ID
Hs.461775
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014427.4 NP_055242.1 17 2636
NM_153636.2 NP_705900.1 15 2411
XM_011523000.1 XP_011521302.1 16 2500
XM_011523001.2 XP_011521303.1 16 2265
XM_017023138.1 XP_016878627.1 18 2694
XM_017023139.1 XP_016878628.1 17 2611
XM_017023140.1 XP_016878629.1 16 2480
XM_017023141.1 XP_016878630.1 15 3587
GenBank mRNA AJ133798.1 - 17 2635
BC035334.1 - 15 2331 5191900
BC064577.1 - 15 2388 5727324

View Full Product Details