Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098531.2 | NP_001092001.1 | 6 | 1086 | |
NM_001098532.2 | NP_001092002.1 | 5 | 850 | ||
NM_006105.5 | NP_006096.2 | 6 | 620 | ||
XM_005268571.3 | XP_005268628.1 | 5 | 630 | ||
XM_011537752.1 | XP_011536054.1 | 6 | 1103 | ||
XM_011537755.1 | XP_011536057.1 | 6 | 665 | ||
XM_011537758.2 | XP_011536060.1 | 5, 6 | 715 | ||
XM_017018688.1 | XP_016874177.1 | 6 | 1103 | ||
GenBank mRNA | AF103905.1 | - | 5 | 410 | |
AK092448.1 | - | 6 | 1058 | ||
AK290230.1 | - | 7 | 751 | ||
AK299029.1 | - | 6 | 714 | ||
AK308079.1 | - | 6 | 688 | ||
BC017728.1 | - | 6 | 700 | 4414360 | |
BC092404.1 | - | 6 | 689 | 6181000 | |
U78168.1 | - | 6 | 625 | ||
U78169.1 | - | 6 | 625 |