Gene Transcripts

Gene Symbol
EVC
Entrez Gene ID
2121
Gene Name
EvC ciliary complex subunit 1
Gene Aliases
DWF-1, EVC1, EVCL
Chromosome Location
Chr. 4: 5711197 - 5829043 on Build GRCh38
UniGene ID
Hs.646899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001306090.1 NP_001293019.1 2 443
NM_001306092.1 NP_001293021.1 2 443
NM_153717.2 NP_714928.1 2 443
XM_006713865.3 XP_006713928.1 2 440
XM_006713866.3 XP_006713929.1 2 440
XM_011513419.2 XP_011511721.1 2 437
XM_017007883.1 XP_016863372.1 2 437
GenBank mRNA AF216184.1 - 2 443
AF216185.1 - 2 443
AF239742.1 - 2 259
BC085608.1 - 2 425 30523373

View Full Product Details