Gene Transcripts

Gene Symbol
ZNF295-AS1
Entrez Gene ID
150142
Gene Name
ZNF295 antisense RNA 1
Gene Aliases
C21orf121, NCRNA00318, PRED87
Chromosome Location
Chr. 21: 42009194 - 42024920 on Build GRCh38
UniGene ID
Hs.143833
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027273.2 2 581
NR_119384.1 2 810
NR_119385.1 2 627
GenBank mRNA AK055167.1 - 1 2641
BC029588.1 - 2 581

View Full Product Details