Gene Transcripts

Gene Symbol
OR5AR1
Entrez Gene ID
219493
Gene Name
olfactory receptor family 5 subfamily AR member 1 (gene/pseudogene)
Gene Aliases
OR11-209
Chromosome Location
Chr. 11: 56663686 - 56664618 on Build GRCh38
UniGene ID
Hs.553633
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004730.1 NP_001004730.1 1 705
GenBank mRNA BC140745.1 - 1 729

View Full Product Details