Gene Transcripts

Gene Symbol
OR13H1
Entrez Gene ID
347468
Gene Name
olfactory receptor family 13 subfamily H member 1
Gene Aliases
ORX1
Chromosome Location
Chr. X: 131544074 - 131545000 on Build GRCh38
UniGene ID
Hs.553711
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004486.1 NP_001004486.1 1 768
GenBank mRNA BC137050.1 - 1 866

View Full Product Details