Gene Transcripts

Gene Symbol
GSX2
Entrez Gene ID
170825
Gene Name
GS homeobox 2
Gene Aliases
GSH2
Chromosome Location
Chr. 4: 54100081 - 54101955 on Build GRCh38
UniGene ID
Hs.371899
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133267.2 NP_573574.1 1 296
GenBank mRNA AB028838.1 - 1 346
BC075089.2 - 1 54 30915389
BC075090.2 - 1 54 30915601
EU596451.1 - 1 296

View Full Product Details