Gene Transcripts

Gene Symbol
PTRH1
Entrez Gene ID
138428
Gene Name
peptidyl-tRNA hydrolase 1 homolog
Gene Aliases
C9orf115, PTH1
Chromosome Location
Chr. 9: 127695024 - 127716714 on Build GRCh38
UniGene ID
Hs.653265
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002913.1 NP_001002913.1 4 451
XM_006716955.3 XP_006717018.1 4 1508
XM_017014276.1 XP_016869765.1 4 1508
XM_017014277.1 XP_016869766.1 4 1508
XM_017014278.1 XP_016869767.1 3 1408
GenBank mRNA AK090922.1 - 5 1890
BC047012.1 - 4 451 5001221

View Full Product Details