Gene Transcripts

Gene Symbol
INCA1
Entrez Gene ID
388324
Gene Name
inhibitor of CDK, cyclin A1 interacting protein 1
Gene Aliases
HSD45
Chromosome Location
Chr. 17: 4988130 - 4997933 on Build GRCh38
UniGene ID
Hs.658926
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167985.1 NP_001161457.1 7 747
NM_001167986.1 NP_001161458.1 8 1079
NM_001167987.1 NP_001161459.1 7 792
NM_213726.2 NP_998891.2 7 909
XM_005256627.3 XP_005256684.1 6 987
XM_005256628.4 XP_005256685.1 6 1184
XM_005256629.1 XP_005256686.1 6 994
XM_006721517.1 XP_006721580.1 6 1039
XM_006721518.2 XP_006721581.1 6 943
XM_006721519.3 XP_006721582.1 6 1140
XM_011523832.2 XP_011522134.1 7 1309
GenBank mRNA AY601906.1 - 7 909
AY601907.1 - 7 747
AY604176.1 - 5 467
BC119781.1 - 7 868 40109268
BC122873.1 - 6 581 40109270

View Full Product Details