Gene Transcripts
- Gene Symbol
- ANKRD20A11P
- Entrez Gene ID
- 391267
- Gene Name
- ankyrin repeat domain 20 family member A11, pseudogene
- Gene Aliases
- C21orf81
- Chromosome Location
- Chr. 21: 13943769 - 13980444 on Build GRCh38
- UniGene ID
- Hs.364456
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027270.1 |
|
6 |
1038 |
|
GenBank mRNA |
AF426257.1 |
- |
6 |
1038 |
|
- Gene Symbol
- ANKRD20A9P
- Entrez Gene ID
- 284232
- Gene Name
- ankyrin repeat domain 20 family member A9, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 13: 18834403 - 18871969 on Build GRCh38
- UniGene ID
- Hs.679496
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027995.1 |
|
1 |
27402 |
|
View Full Product Details