Gene Transcripts

Gene Symbol
HOPX
Entrez Gene ID
84525
Gene Name
HOP homeobox
Gene Aliases
CAMEO, HOD, HOP, LAGY, NECC1, OB1, SMAP31, TOTO
Chromosome Location
Chr. 4: 56647988 - 56681866 on Build GRCh38
UniGene ID
Hs.619396
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145459.1 NP_001138931.1 3 841
NM_001145460.1 NP_001138932.1 5 1325
NM_032495.5 NP_115884.4 4 1195
NM_139211.4 NP_631957.1 3 797
NM_139212.3 NP_631958.1 3 1070
XM_017008728.1 XP_016864217.1 3 832
XM_017008729.1 XP_016864218.1 3 714
XM_017008730.1 XP_016864219.1 3 762
XM_017008731.1 XP_016864220.1 3 797
XM_017008732.1 XP_016864221.1 4 1073
XM_017008733.1 XP_016864222.1 3 637
XM_017008734.1 XP_016864223.1 3 693
GenBank mRNA AB059408.1 - 3 755
AB059409.1 - 3 638
AB059410.2 - 3 647
AF454763.1 - 3 661
AF492675.1 - 3 1045
AF492676.1 - 4 777
AF492677.1 - 4 821
AF492678.1 - 5 951
AF492679.1 - 5 907
AF492680.1 - 3 797
AF492681.1 - 3 760
AK123318.1 - 1 2867
BC014225.2 - 2 910 4335211
EB386856.1 - 3 791

View Full Product Details