Gene Transcripts
- Gene Symbol
- PLEKHM1
- Entrez Gene ID
- 9842
- Gene Name
- pleckstrin homology and RUN domain containing M1
- Gene Aliases
- AP162, B2, OPTB6
- Chromosome Location
- Chr. 17: 45434262 - 45490780 on Build GRCh38
- UniGene ID
- Hs.514242
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014798.2 |
NP_055613.1
|
9, 10 |
3015 |
|
NR_027774.1 |
|
8, 9 |
2767 |
|
NR_027782.1 |
|
8, 9 |
2630 |
|
XM_006722201.3 |
XP_006722264.1
|
7, 8 |
2796 |
|
XM_011525523.2 |
XP_011523825.1
|
9, 10 |
2986 |
|
XM_011525524.1 |
XP_011523826.1
|
9, 10 |
3165 |
|
XM_011525525.1 |
XP_011523827.1
|
8, 9 |
2773 |
|
XM_011525526.1 |
XP_011523828.1
|
10, 11 |
2809 |
|
XM_011525528.2 |
XP_011523830.1
|
4, 5 |
1359 |
|
XM_017025451.1 |
XP_016880940.1
|
9, 10 |
2963 |
|
XM_017025452.1 |
XP_016880941.1
|
8, 9 |
2718 |
|
XM_017025453.1 |
XP_016880942.1
|
10, 11 |
3172 |
|
XM_017025454.1 |
XP_016880943.1
|
8, 9 |
2863 |
|
GenBank mRNA |
AB002354.2 |
- |
9, 10 |
2968 |
|
AJ002220.1 |
- |
7, 8 |
2603 |
|
AK074103.1 |
- |
5, 6 |
1763 |
|
AK299116.1 |
- |
8, 9 |
2715 |
|
AK299467.1 |
- |
8, 9 |
2769 |
|
BC064361.1 |
- |
9, 10 |
2979 |
5535600 |
View Full Product Details