Gene Transcripts

Gene Symbol
Dnajb13
Entrez Gene ID
308857
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 13
Gene Aliases
Tsarg1
Chromosome Location
Chr. 1: 165515718 - 165530163 on Build Rnor_6.0
UniGene ID
Rn.45046
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005885.2 NP_001005885.1 8 950
XM_006229766.2 XP_006229828.1 8 1015
GenBank mRNA AY325765.1 - 8 990
AY380804.1 - 8 1008
BC098002.1 - 8 950 7453525

View Full Product Details