Gene Transcripts

Gene Symbol
Ifit3
Entrez Gene ID
309526
Gene Name
interferon-induced protein with tetratricopeptide repeats 3
Gene Aliases
Chromosome Location
Chr. 1: 252906234 - 252911377 on Build Rnor_6.0
UniGene ID
Rn.35775
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007694.1 NP_001007695.1 2 303
GenBank mRNA BC079079.1 - 2 303

View Full Product Details