Gene Transcripts

Gene Symbol
RGD1359634
Entrez Gene ID
315126
Gene Name
similar to RIKEN cDNA 1700088E04
Gene Aliases
Chromosome Location
Chr. 7: 120372874 - 120380463 on Build Rnor_6.0
UniGene ID
Rn.24486
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001007708.1 NP_001007709.1 1, 2 356
XM_006241992.2 XP_006242054.1 2, 3 203
GenBank mRNA BC079039.1 - 1, 2 356

View Full Product Details