Gene Transcripts
- Gene Symbol
- LINC00909
- Entrez Gene ID
- 400657
- Gene Name
- long intergenic non-protein coding RNA 909
- Gene Aliases
- Chromosome Location
- Chr. 18: 74591774 - 74597835 on Build GRCh38
- UniGene ID
- Hs.61508
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024484.1 |
|
2 |
2285 |
|
GenBank mRNA |
AK001853.1 |
- |
2 |
2032 |
|
BC036588.2 |
- |
2 |
2276 |
|
View Full Product Details