Gene Transcripts

Gene Symbol
RGD1306954
Entrez Gene ID
288269
Gene Name
similar to RIKEN cDNA 1110004E09
Gene Aliases
Chromosome Location
Chr. 11: 31094103 - 31103487 on Build Rnor_6.0
UniGene ID
Rn.24386
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008288.1 NP_001008289.1 3, 4 475
GenBank mRNA BC085340.1 - 3, 4 475

View Full Product Details