Gene Transcripts

Gene Symbol
Mkrn2
Entrez Gene ID
297525
Gene Name
makorin, ring finger protein, 2
Gene Aliases
Chromosome Location
Chr. 4: 147514120 - 147532086 on Build Rnor_6.0
UniGene ID
Rn.129070
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008314.1 NP_001008315.1 8 1432
GenBank mRNA BC083870.1 - 8 1432

View Full Product Details