Gene Transcripts

Gene Symbol
Pmvk
Entrez Gene ID
310645
Gene Name
phosphomevalonate kinase
Gene Aliases
Chromosome Location
Chr. 2: 188784329 - 188793885 on Build Rnor_6.0
UniGene ID
Rn.22202
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008352.1 NP_001008353.1 5 950
GenBank mRNA BC086349.1 - 5 950
FQ226470.1 - 5 1080
FQ233923.1 - 5 1080
FQ234368.1 - 5 1080

View Full Product Details