Gene Transcripts

Gene Symbol
TCHHL1
Entrez Gene ID
126637
Gene Name
trichohyalin like 1
Gene Aliases
S100A17, THHL1
Chromosome Location
Chr. 1: 152084144 - 152089064 on Build GRCh38
UniGene ID
Hs.546477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008536.1 NP_001008536.1 3 1354
GenBank mRNA AY456639.1 - 3 1354
BC137493.1 - 3 1344 9021493

View Full Product Details