Gene Transcripts

Gene Symbol
Whrn
Entrez Gene ID
73750
Gene Name
whirlin
Gene Aliases
1110035G07Rik, AW122018, AW742671, C430046P22Rik, Dfnb31, wi
Chromosome Location
Chr. 4: 63414909 - 63496078 on Build GRCm38
UniGene ID
Mm.300397
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008791.2 NP_001008791.1 13 3802
NM_001008792.2 NP_001008792.1 12 3781
NM_001008793.2 NP_001008793.1 12 3766
NM_028640.2 NP_082916.1 12 3769
GenBank mRNA AK004110.1 - 2 499
AK122523.1 - 12 2897
AY739114.1 - 12 3729
AY739115.1 - 13 3762
AY739116.1 - 12 3741
AY739117.1 - 12 3726
AY739118.1 - 9 2378
AY739119.1 - 8 2345
AY739120.1 - 7 2132
AY739121.1 - 7 2250
AY739122.1 - 8 2582

View Full Product Details