Gene Transcripts

Gene Symbol
Vom1r57
Entrez Gene ID
494296
Gene Name
vomeronasal 1 receptor 57
Gene Aliases
V1rk1
Chromosome Location
Chr. 1: 66687478 - 66688422 on Build Rnor_6.0
UniGene ID
Rn.129411
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008951.1 NP_001008951.1 1 178
GenBank mRNA AY510278.1 - 1 178

View Full Product Details