Gene Transcripts

Gene Symbol
CDKL4
Entrez Gene ID
344387
Gene Name
cyclin dependent kinase like 4
Gene Aliases
Chromosome Location
Chr. 2: 39174421 - 39246815 on Build GRCh38
UniGene ID
Hs.403201
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009565.1 NP_001009565.1 8 935
GenBank mRNA AY845084.1 - 8 935
BC144460.1 - 9 1108

View Full Product Details