Gene Transcripts

Gene Symbol
LRRC27
Entrez Gene ID
80313
Gene Name
leucine rich repeat containing 27
Gene Aliases
Chromosome Location
Chr. 10: 132330063 - 132381506 on Build GRCh38
UniGene ID
Hs.119897
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143757.1 NP_001137229.1 6 1210
NM_001143758.1 NP_001137230.1 6 1057
NM_001143759.1 NP_001137231.1 6 1057
NM_001309474.1 NP_001296403.1 5 967
NM_030626.2 NP_085129.1 6 1057
XM_006717983.2 XP_006718046.1 6 1370
XM_006717984.2 XP_006718047.1 6 1057
XM_011540208.1 XP_011538510.1 6 1140
XM_011540209.1 XP_011538511.1 6 1057
XM_011540210.2 XP_011538512.1 5 965
XM_017016690.1 XP_016872179.1 6 1057
XM_017016691.1 XP_016872180.1 6 1057
XM_017016692.1 XP_016872181.1 6 1057
XM_017016693.1 XP_016872182.1 6 1057
XM_017016694.1 XP_016872183.1 5 944
GenBank mRNA AB051461.1 - 6 967
AK093250.1 - 6 823
AK097418.1 - 6 1168
AK098652.1 - 6 1013
AK301422.1 - 5 734
BC050097.1 - 6 1012 5273395

View Full Product Details