Gene Transcripts

Gene Symbol
Ajuba
Entrez Gene ID
85265
Gene Name
ajuba LIM protein
Gene Aliases
Jub
Chromosome Location
Chr. 15: 33208210 - 33218456 on Build Rnor_6.0
UniGene ID
Rn.144617
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053503.1 NP_445955.1 8 2738
GenBank mRNA BC085802.1 - 8 2738

View Full Product Details