Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109911.1 | NP_001103381.1 | 14 | 1415 | |
NM_053876.2 | NP_446328.2 | 15 | 1298 | ||
XM_006241116.2 | XP_006241178.1 | 14 | 2605 | ||
XM_008765198.1 | XP_008763420.1 | 12 | 1145 | ||
GenBank mRNA | AY724543.1 | - | 14 | 1402 | |
BC088317.1 | - | 10 | 926 | 7325202 | |
U32577.2 | - | 15 | 1298 |