Gene Transcripts

Gene Symbol
BTF3L4
Entrez Gene ID
91408
Gene Name
basic transcription factor 3 like 4
Gene Aliases
Chromosome Location
Chr. 1: 52056185 - 52090716 on Build GRCh38
UniGene ID
Hs.429839
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136497.2 NP_001129969.1 5 672
NM_001243767.1 NP_001230696.1 5 537
NM_152265.4 NP_689478.1 6 739
GenBank mRNA AK027582.1 - 5 553
AK027750.1 - 6 621
BC021004.1 - 5 516
BC022371.1 - 6 601 4739327
BC070378.1 - 6 629 6495112
BU785869.1 - 5 381
DA369498.1 - 5 458

View Full Product Details