Gene Transcripts

Gene Symbol
Bace2
Entrez Gene ID
288227
Gene Name
beta-site APP-cleaving enzyme 2
Gene Aliases
Chromosome Location
Chr. 11: 37798397 - 37882076 on Build Rnor_6.0
UniGene ID
Rn.38581
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002802.1 NP_001002802.1 9 1437
XM_006248153.2 XP_006248215.1 9 1491
GenBank mRNA BC166597.1 - 9 1451

View Full Product Details