Gene Transcripts

Gene Symbol
Ddx56
Entrez Gene ID
289780
Gene Name
DEAD (Asp-Glu-Ala-Asp) box helicase 56
Gene Aliases
Ddx21, RH-II/Gu
Chromosome Location
Chr. 14: 86379010 - 86387623 on Build Rnor_6.0
UniGene ID
Rn.39458
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004211.1 NP_001004211.1 5 806
XM_006251437.2 XP_006251499.1 6 741
GenBank mRNA BC078919.1 - 5 806
FQ212430.1 - 727

View Full Product Details