Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001004239.1 | NP_001004239.1 | 11 | 1027 | |
XM_006241448.1 | XP_006241510.1 | 12 | 1115 | ||
XM_008765392.1 | XP_008763614.1 | 12 | 1117 | ||
GenBank mRNA | BC079042.1 | - | 11 | 1027 |