Gene Transcripts

Gene Symbol
Nuak1
Entrez Gene ID
77976
Gene Name
NUAK family, SNF1-like kinase, 1
Gene Aliases
AU014801, AW494241, B230104P22Rik, Omphk1
Chromosome Location
Chr. 10: 84370900 - 84440471 on Build GRCm38
UniGene ID
Mm.25874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004363.1 NP_001004363.1 7 3912
GenBank mRNA AB182364.1 - 6 3549
BC040467.1 - 3 3208
BC082328.1 - 7 3912 30355405

View Full Product Details